Product Details

SNP ID
rs142645437
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:88753498 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAATGAGCCTCCTAATGACTGTAG[C/T]ACAGACATAGAGTCTCCATCTGCTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CPXCR1 PubMed Links

Gene Details

Gene
CPXCR1
Gene Name
CPX chromosome region, candidate 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184771.1 343 Silent Mutation AGC,AGT S28S NP_001171700.1
NM_033048.5 343 Silent Mutation AGC,AGT S28S NP_149037.4

View Full Product Details