Product Details

SNP ID
rs144063480
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:103253659 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAAGCACACAGGATAAGGACGGCT[A/G]CGTGAATGTCTTTGCTTCCAATGCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TCEAL8 PubMed Links

Gene Details

Gene
TCEAL8
Gene Name
transcription elongation factor A like 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001006684.1 475 Silent Mutation NP_001006685.1
NM_153333.2 475 Silent Mutation NP_699164.1

View Full Product Details