Product Details

SNP ID
rs144735761
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:53430547 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGCTCCCTACCGGGTCCTGCCCT[A/T]TTGCTGGAAGGGCATGACTCCAGAG
Phenotype
MIM: 300256 MIM: 300040
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
HSD17B10 PubMed Links

Gene Details

Gene
HSD17B10
Gene Name
hydroxysteroid 17-beta dehydrogenase 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037811.2 1053 Intron NP_001032900.1
NM_004493.2 1053 Intron NP_004484.1
Gene
RIBC1
Gene Name
RIB43A domain with coiled-coils 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031745.4 1053 Missense Mutation TAT,TTT Y272F NP_001026915.1
NM_001267053.3 1053 Missense Mutation TAT,TTT Y157F NP_001253982.1
NM_144968.3 1053 Intron NP_659405.1
XM_005261988.3 1053 Missense Mutation TAT,TTT Y272F XP_005262045.1
XM_005261990.3 1053 Missense Mutation TAT,TTT Y157F XP_005262047.1
Gene
SMC1A
Gene Name
structural maintenance of chromosomes 1A
There are no transcripts associated with this gene.

View Full Product Details