Product Details

SNP ID
rs144775134
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:30559630 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCTACACTTGTCATGATGAGTTC[A/G]GACGCCAGAATCTGGGTAGGAGAAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CXorf21 PubMed Links

Gene Details

Gene
CXorf21
Gene Name
chromosome X open reading frame 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025159.2 1121 Silent Mutation TCC,TCT S242S NP_079435.1

View Full Product Details