Product Details

SNP ID
rs144899652
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:44844694 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTATGAGAAGGACCTACGTATGATG[A/G]TATCAATGTAAGCCATCCCGGCCAG
Phenotype
MIM: 300678
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DUSP21 PubMed Links

Gene Details

Gene
DUSP21
Gene Name
dual specificity phosphatase 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022076.3 692 Missense Mutation ATA,GTA I188V NP_071359.3

View Full Product Details