Product Details

SNP ID
rs145647521
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:119871988 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCCAGGACTGGCTACTGCGTACA[C/T]CCACAGGTTCACTAACGGGGGCAAG
Phenotype
MIM: 300078 MIM: 300951
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NDUFA1 PubMed Links

Gene Details

Gene
NDUFA1
Gene Name
NADH:ubiquinone oxidoreductase subunit A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004541.3 218 Missense Mutation ACC,ATC T26I NP_004532.1
Gene
RNF113A
Gene Name
ring finger protein 113A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006978.2 218 Intron NP_008909.1

View Full Product Details