Product Details

SNP ID
rs145726572
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:18174066 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGGTTCCTCTGGATTATCAGTGG[A/G]AGGTTCTCGCATTCCTTCAGGGATA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BEND2 PubMed Links

Gene Details

Gene
BEND2
Gene Name
BEN domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184767.1 1826 Missense Mutation CCC,TCC P558S NP_001171696.1
NM_153346.4 1826 Missense Mutation CCC,TCC P649S NP_699177.2
XM_011545447.1 1826 Missense Mutation CCC,TCC P663S XP_011543749.1
XM_017029266.1 1826 Missense Mutation CCC,TCC P621S XP_016884755.1
XM_017029267.1 1826 Missense Mutation CCC,TCC P663S XP_016884756.1

View Full Product Details