Product Details

SNP ID
rs145766767
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:34944353 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGGCAAACGAGTGTGCTTCAAGG[C/T]TGATGTACGGCATGGAGCTAGACGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM47B PubMed Links

Gene Details

Gene
FAM47B
Gene Name
family with sequence similarity 47 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152631.2 Intron NP_689844.2

View Full Product Details