Product Details

SNP ID
rs146663704
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:120254085 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGGTCCAATCTAACCCAGGCCCT[A/G]TTGCTATTTCAGATGTTGCACCTAG
Phenotype
MIM: 300329
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NKAPP1 PubMed Links

Gene Details

Gene
NKAPP1
Gene Name
NFKB activating protein pseudogene 1
There are no transcripts associated with this gene.

Gene
TMEM255A
Gene Name
transmembrane protein 255A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001104544.1 997 Intron NP_001098014.1
NM_001104545.1 997 Intron NP_001098015.1
NM_017938.3 997 Intron NP_060408.3
XM_017029619.1 997 Intron XP_016885108.1
Gene
ZBTB33
Gene Name
zinc finger and BTB domain containing 33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184742.1 997 Missense Mutation ATT,GTT I224V NP_001171671.1
NM_006777.3 997 Missense Mutation ATT,GTT I224V NP_006768.1

View Full Product Details