Product Details

SNP ID
rs146885760
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.X:73213331 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATTGTGACCAAGTAAAAAATCAT[A/C]ATTTCCATCCCTTCCATTTGAGGTG
Phenotype
MIM: 300026
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
NAP1L2 PubMed Links

Gene Details

Gene
NAP1L2
Gene Name
nucleosome assembly protein 1 like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021963.3 1544 Missense Mutation GAT,TAT D388Y NP_068798.1

View Full Product Details