Product Details

SNP ID
rs147131196
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:37894941 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTTTTCAGTCCTTTGGATTCTGA[A/C]TGAGTTATACCACCAGCTTTCCTGG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SYTL5 PubMed Links

Gene Details

Gene
SYTL5
Gene Name
synaptotagmin like 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163334.1 Intron NP_001156806.1
NM_001163335.1 Intron NP_001156807.1
NM_138780.2 Intron NP_620135.1
XM_011544001.2 Intron XP_011542303.1
XM_011544002.2 Intron XP_011542304.1
XM_017029972.1 Intron XP_016885461.1
XM_017029973.1 Intron XP_016885462.1
XM_017029974.1 Intron XP_016885463.1

View Full Product Details