Product Details

SNP ID
rs147653963
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:84062264 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATAGAAAACCTGAAAGTTATTTT[A/G]TAATCATTTTTCATTTTCTGATTTC
Phenotype
MIM: 300303
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RPS6KA6 PubMed Links

Gene Details

Gene
RPS6KA6
Gene Name
ribosomal protein S6 kinase A6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014496.4 4287 UTR 3 NP_055311.1
XM_011530917.2 4287 Intron XP_011529219.1
XM_011530918.2 4287 Intron XP_011529220.1
XM_011530919.2 4287 Intron XP_011529221.1
XM_011530920.2 4287 Intron XP_011529222.1
XM_017029423.1 4287 UTR 3 XP_016884912.1
XM_017029424.1 4287 Intron XP_016884913.1
XM_017029425.1 4287 Intron XP_016884914.1
XM_017029426.1 4287 Intron XP_016884915.1

View Full Product Details