Product Details

SNP ID
rs147687583
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:137030182 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTAGCAGAATCGTAGGAAGGGACA[A/C]TCTTGCCTTCAGTCCCACCCTCTGT
Phenotype
MIM: 300393
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GPR101 PubMed Links

Gene Details

Gene
GPR101
Gene Name
G protein-coupled receptor 101
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_054021.1 1493 Missense Mutation NP_473362.1

View Full Product Details