Product Details

SNP ID
rs148223311
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:79171796 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTGAAGTCCAATGAAATTAAAAG[C/T]TGCCTAGCCAGAAGGGTGATTCTAA
Phenotype
MIM: 300903
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPR174 PubMed Links

Gene Details

Gene
GPR174
Gene Name
G protein-coupled receptor 174
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032553.1 825 Silent Mutation AGC,AGT S263S NP_115942.1

View Full Product Details