Product Details

SNP ID
rs148315361
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:44844457 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGTACGCTGCTGCACTGCATGGCT[G/T]GAGTGAGCCGTTCCGCCTCACTGTG
Phenotype
MIM: 300678
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DUSP21 PubMed Links

Gene Details

Gene
DUSP21
Gene Name
dual specificity phosphatase 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022076.3 455 Nonsense Mutation GGA,TGA G109* NP_071359.3

View Full Product Details