Product Details

SNP ID
rs148435716
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:34942842 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGAGAGGGCCACCATGGGGGACC[A/G]GAGGCCACAGGACCGGCCAAGGTCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM47B PubMed Links

Gene Details

Gene
FAM47B
Gene Name
family with sequence similarity 47 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152631.2 Intron NP_689844.2

View Full Product Details