Product Details

SNP ID
rs148668161
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:153057573 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGTGTTTTCTGGGAACACCATAT[C/G]CATCCCAGGTGCACTGGCCTTTGAT
Phenotype
MIM: 300675
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PNMA3 PubMed Links

Gene Details

Gene
PNMA3
Gene Name
paraneoplastic Ma antigen 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282535.1 854 Missense Mutation TCC,TGC S173C NP_001269464.1
NM_013364.5 854 Missense Mutation TCC,TGC S173C NP_037496.4

View Full Product Details