Product Details

SNP ID
rs148964045
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:93671150 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACACCTCAGTGGTAAACAATAACA[C/T]CATTATGATTAAATTTATAGGCCAC
Phenotype
MIM: 300117
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM133A PubMed Links

Gene Details

Gene
FAM133A
Gene Name
family with sequence similarity 133 member A
There are no transcripts associated with this gene.

Gene
NAP1L3
Gene Name
nucleosome assembly protein 1 like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004538.5 2534 UTR 3 NP_004529.2

View Full Product Details