Product Details

SNP ID
rs149192322
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:106033693 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTTCACCAATGTGCAGCACAGCC[A/T]TATGGGCAGCCTGTGTGGGGAGAAC
Phenotype
MIM: 314200
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SERPINA7 PubMed Links

Gene Details

Gene
SERPINA7
Gene Name
serpin family A member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000354.5 1102 Missense Mutation AAG,ATG K352M NP_000345.2
XM_006724683.2 1102 Missense Mutation AAG,ATG K362M XP_006724746.1

View Full Product Details