Product Details
- SNP ID
-
rs149758385
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:66599684 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGCAGGAGGCCATGGTAAAGGACT[A/C]CTGTGTGGGGAAGCCACTAGTCGAG
- Phenotype
-
MIM: 300276
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
EDA2R
PubMed Links
Gene Details
- Gene
- EDA2R
- Gene Name
- ectodysplasin A2 receptor
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001199687.2 |
865 |
Nonsense Mutation |
GAG,TAG |
E232* |
NP_001186616.1 |
NM_001242310.1 |
865 |
Nonsense Mutation |
GAG,TAG |
E253* |
NP_001229239.1 |
NM_001324199.1 |
865 |
Nonsense Mutation |
GAG,TAG |
E108* |
NP_001311128.1 |
NM_001324201.1 |
865 |
Nonsense Mutation |
GAG,TAG |
E200* |
NP_001311130.1 |
NM_001324202.1 |
865 |
Intron |
|
|
NP_001311131.1 |
NM_001324204.1 |
865 |
Intron |
|
|
NP_001311133.1 |
NM_001324205.1 |
865 |
Intron |
|
|
NP_001311134.1 |
NM_001324206.1 |
865 |
Nonsense Mutation |
GAG,TAG |
E234* |
NP_001311135.1 |
NM_021783.4 |
865 |
Nonsense Mutation |
GAG,TAG |
E232* |
NP_068555.1 |
XM_011531000.1 |
865 |
Nonsense Mutation |
GAG,TAG |
E253* |
XP_011529302.1 |
XM_011531001.1 |
865 |
Nonsense Mutation |
GAG,TAG |
E253* |
XP_011529303.1 |
XM_011531002.1 |
865 |
Intron |
|
|
XP_011529304.1 |
XM_011531003.2 |
865 |
Nonsense Mutation |
GAG,TAG |
E173* |
XP_011529305.1 |
XM_011531004.1 |
865 |
Intron |
|
|
XP_011529306.1 |
XM_011531005.2 |
865 |
Nonsense Mutation |
GAG,TAG |
E108* |
XP_011529307.1 |
XM_011531006.2 |
865 |
Intron |
|
|
XP_011529308.1 |
XM_011531007.2 |
865 |
Intron |
|
|
XP_011529309.1 |
XM_017029703.1 |
865 |
Nonsense Mutation |
GAG,TAG |
E200* |
XP_016885192.1 |
XM_017029704.1 |
865 |
Intron |
|
|
XP_016885193.1 |
XM_017029705.1 |
865 |
Nonsense Mutation |
GAG,TAG |
E154* |
XP_016885194.1 |
XM_017029706.1 |
865 |
Intron |
|
|
XP_016885195.1 |
XM_017029707.1 |
865 |
Nonsense Mutation |
GAG,TAG |
E129* |
XP_016885196.1 |
XM_017029708.1 |
865 |
Intron |
|
|
XP_016885197.1 |
View Full Product Details