Product Details

SNP ID
rs150331463
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:137030473 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACATAGGAGAAAATGATGATGAAG[A/G]TCACTTTAGCAGCTTTGCACTGGTA
Phenotype
MIM: 300393
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPR101 PubMed Links

Gene Details

Gene
GPR101
Gene Name
G protein-coupled receptor 101
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_054021.1 1202 Missense Mutation ACC,ATC T401I NP_473362.1

View Full Product Details