Product Details

SNP ID
rs150401054
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:37695029 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGATCCAGGCTTGGGAGTTGTGG[G/T]ACATCACTTCTGCTTCCTTCTCATT
Phenotype
MIM: 314850
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
XK PubMed Links

Gene Details

Gene
XK
Gene Name
X-linked Kx blood group
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011543978.2 Intron XP_011542280.1

View Full Product Details