Product Details

SNP ID
rs150572239
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:46930208 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCTTACTCTCCATTCTGAGGCAA[C/T]CATGTGAGGAAGCCCCAGCTAACCT
Phenotype
MIM: 300618
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
JADE3 PubMed Links

Gene Details

Gene
JADE3
Gene Name
jade family PHD finger 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077445.2 Intron NP_001070913.1
NM_014735.4 Intron NP_055550.1

View Full Product Details