Product Details
- SNP ID
-
rs150879214
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:2854993 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTTCTGCGTGTTTTGCTTCACCAG[A/G]TGCTGTCCAATGTCGATGAGCTGTT
- Phenotype
-
MIM: 300198
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GYG2
PubMed Links
Gene Details
- Gene
- GYG2
- Gene Name
- glycogenin 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001079855.1 |
700 |
Missense Mutation |
ATG,GTG |
M109V |
NP_001073324.1 |
NM_001184702.1 |
700 |
Missense Mutation |
ATG,GTG |
M109V |
NP_001171631.1 |
NM_001184703.1 |
700 |
Missense Mutation |
ATG,GTG |
M140V |
NP_001171632.1 |
NM_001184704.1 |
700 |
UTR 5 |
|
|
NP_001171633.1 |
NM_003918.2 |
700 |
Missense Mutation |
ATG,GTG |
M140V |
NP_003909.2 |
XM_011545600.2 |
700 |
Missense Mutation |
ATG,GTG |
M109V |
XP_011543902.1 |
XM_017029927.1 |
700 |
Missense Mutation |
ATG,GTG |
M140V |
XP_016885416.1 |
XM_017029928.1 |
700 |
Missense Mutation |
ATG,GTG |
M140V |
XP_016885417.1 |
XM_017029929.1 |
700 |
Missense Mutation |
ATG,GTG |
M140V |
XP_016885418.1 |
XM_017029930.1 |
700 |
Missense Mutation |
ATG,GTG |
M109V |
XP_016885419.1 |
XM_017029931.1 |
700 |
Missense Mutation |
ATG,GTG |
M109V |
XP_016885420.1 |
View Full Product Details