Product Details

SNP ID
rs151043660
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:154488101 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTCATGGAGATGGCCAGGGTGAC[A/G]TCCCCTCCAAGAAGGAGGCTGAAGA
Phenotype
MIM: 300060 MIM: 312090 MIM: 312070
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LAGE3 PubMed Links

Gene Details

Gene
LAGE3
Gene Name
L antigen family member 3
There are no transcripts associated with this gene.

Gene
SLC10A3
Gene Name
solute carrier family 10 member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142391.2 1222 Silent Mutation GAC,GAT D251D NP_001135863.1
NM_001142392.2 1222 Silent Mutation GAC,GAT D280D NP_001135864.1
NM_019848.4 1222 Silent Mutation GAC,GAT D280D NP_062822.1
XM_005277913.4 1222 Silent Mutation GAC,GAT D335D XP_005277970.1
XM_006724847.3 1222 Silent Mutation GAC,GAT D373D XP_006724910.1
XM_006724848.3 1222 Silent Mutation GAC,GAT D344D XP_006724911.1
XM_011531200.2 1222 Silent Mutation GAC,GAT D335D XP_011529502.2
XM_011531201.2 1222 Silent Mutation GAC,GAT D280D XP_011529503.1
Gene
UBL4A
Gene Name
ubiquitin like 4A
There are no transcripts associated with this gene.

View Full Product Details