Product Details

SNP ID
rs1049274
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:28201322 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTCTTCTCTTTCTTCCCCTTCG[T/G]ATTGGCTTGGAAGTTTCGCCAGCTG
Phenotype
MIM: 173393
Polymorphism
T/G, Transversion substitution
Allele Nomenclature
Literature Links
DNAJC8 PubMed Links

Gene Details

Gene
DNAJC8
Gene Name
DnaJ heat shock protein family (Hsp40) member C8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014280.2 721 Missense Mutation ACG,CCG T230P NP_055095.2
Gene
PTAFR
Gene Name
platelet activating factor receptor
There are no transcripts associated with this gene.

View Full Product Details