Product Details

SNP ID
rs1127958
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:150801721 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGCAGCCGGCTGCTGGTGGCCTC[A/G]TGGGTGAGTGTGACGGCCTGCCTCG
Phenotype
MIM: 610334 MIM: 610385
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM176A PubMed Links

Gene Details

Gene
TMEM176A
Gene Name
transmembrane protein 176A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018487.2 384 Silent Mutation TCA,TCG S57S NP_060957.2
XM_011516376.2 384 Silent Mutation TCA,TCG S74S XP_011514678.1
XM_011516377.2 384 Silent Mutation TCA,TCG S74S XP_011514679.1
XM_011516378.2 384 Silent Mutation TCA,TCG S74S XP_011514680.1
XM_017012393.1 384 Silent Mutation TCA,TCG S57S XP_016867882.1
Gene
TMEM176B
Gene Name
transmembrane protein 176B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001101311.1 384 Intron NP_001094781.1
NM_001101312.1 384 Intron NP_001094782.1
NM_001101314.1 384 Intron NP_001094784.1
NM_014020.3 384 Intron NP_054739.3
XM_006715933.3 384 Intron XP_006715996.2

View Full Product Details