Product Details

SNP ID
rs11465393
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:7743110 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTTCTTCATCCCTGGAGCAGGAGG[C/A]TGCGGACTTTTTGCAGATCCAGAAT
Phenotype
MIM: 604672
Polymorphism
C/A, Transversion substitution
Allele Nomenclature
Literature Links
CD209 PubMed Links

Gene Details

Gene
CD209
Gene Name
CD209 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144893.1 828 Missense Mutation GCC,TCC A246S NP_001138365.1
NM_001144894.1 828 Missense Mutation GCC,TCC A338S NP_001138366.1
NM_001144895.1 828 Missense Mutation GCC,TCC A290S NP_001138367.1
NM_001144896.1 828 Missense Mutation GCC,TCC A358S NP_001138368.1
NM_001144897.1 828 Missense Mutation GCC,TCC A376S NP_001138369.1
NM_001144899.1 828 Missense Mutation GCC,TCC A221S NP_001138371.1
NM_021155.3 828 Missense Mutation GCC,TCC A382S NP_066978.1
XM_005272472.4 828 Missense Mutation GCC,TCC A244S XP_005272529.1

View Full Product Details