Product Details

SNP ID
rs11545989
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:68042276 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGTGGTAGATGGAGGTGCCATCC[C/G]AGCATTCATTTCTCTGTTGGCATCT
Phenotype
MIM: 600685
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KPNA2 PubMed Links

Gene Details

Gene
KPNA2
Gene Name
karyopherin subunit alpha 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320611.1 1114 Missense Mutation CCA,CGA P165R NP_001307540.1
NM_002266.3 1114 Intron NP_002257.1

View Full Product Details