Product Details

SNP ID
rs11551204
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.8:38163773 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCAGCTTGGTCTGCTGTTCCACC[C/T]TTTGTTCTTCTAGAATTTCTTCAAT
Phenotype
MIM: 607281
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
LSM1 PubMed Links

Gene Details

Gene
LSM1
Gene Name
LSM1 homolog, mRNA degradation associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014462.2 709 Missense Mutation AAG,AGG K100R NP_055277.1

View Full Product Details