Product Details

SNP ID
rs11553697
Assay Type
Functionally Tested
NCBI dbSNP Submissions
19
Location
Chr.22:50245986 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCAGGAGTGCAGCGTGGGGGCCC[T/C]GCAGGCCATGGCCAGGCCCCAGCGC
Phenotype
MIM: 608544 MIM: 602399 MIM: 610053
Polymorphism
T/C, Transition Substitution
Allele Nomenclature
Literature Links
HDAC10 PubMed Links

Gene Details

Gene
HDAC10
Gene Name
histone deacetylase 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159286.1 2130 Missense Mutation CAG,CGG Q566R NP_001152758.1
NM_032019.5 2130 Missense Mutation CAG,CGG Q586R NP_114408.3
Gene
MAPK12
Gene Name
mitogen-activated protein kinase 12
There are no transcripts associated with this gene.

Gene
TUBGCP6
Gene Name
tubulin gamma complex associated protein 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020461.3 2130 Intron NP_065194.2

View Full Product Details