Product Details

SNP ID
rs11559096
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:30547255 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAGGGCTCCTGGATCTGGGCGATA[T/G]GGTAGATCCCTGCCAGAACCTGAGG
Phenotype
MIM: 143024
Polymorphism
T/G, Transversion substitution
Allele Nomenclature
Literature Links
GNL1 PubMed Links

Gene Details

Gene
GNL1
Gene Name
G protein nucleolar 1 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005275.3 2689 Missense Mutation CAT,CCT H433P NP_005266.2
XM_005249015.3 2689 Missense Mutation CAT,CCT H433P XP_005249072.1
Gene
PRR3
Gene Name
proline rich 3
There are no transcripts associated with this gene.

View Full Product Details