Product Details
- SNP ID
-
rs1130335
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:232378876 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTACAGCTCTCCCTGGGCATCATCC[C/T]AGGTAATGAGGCTCCCCGAGCTGCC
- Phenotype
-
MIM: 171800
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ALPP
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs10642925] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ALPP
- Gene Name
- alkaline phosphatase, placental
There are no transcripts associated with this gene.
- Gene
- ECEL1P2
- Gene Name
- endothelin converting enzyme like 1 pseudogene 2
There are no transcripts associated with this gene.
View Full Product Details