Product Details

SNP ID
rs7254995
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:20545971 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAAGATCTCGGGCAAAATGAGAAC[A/C]CGTAACTGAAAGAAACAATAAAAGC
Phenotype
MIM: 603984
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZNF737 PubMed Links

Gene Details

Gene
ZNF737
Gene Name
zinc finger protein 737
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159293.1 412 Missense Mutation GGT,TGT G78C NP_001152765.1
XM_005259697.3 412 Missense Mutation GGT,TGT G78C XP_005259754.1
XM_006722597.3 412 Missense Mutation GGT,TGT G78C XP_006722660.1
XM_011527598.2 412 Intron XP_011525900.1
XM_011527599.2 412 Intron XP_011525901.1
XM_017026116.1 412 Intron XP_016881605.1
XM_017026117.1 412 Intron XP_016881606.1

View Full Product Details