Product Details

SNP ID
rs1004665
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24425353 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAAGCTCCCTGGCTTGGTGAGCCC[A/G]GGTGACTAAATTCTACTGAGACACA
Phenotype
MIM: 612978
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CBLN3 PubMed Links

Gene Details

Gene
CBLN3
Gene Name
cerebellin 3 precursor
There are no transcripts associated with this gene.

Gene
KHNYN
Gene Name
KH and NYN domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290256.1 Intron NP_001277185.1
NM_001290257.1 Intron NP_001277186.1
NM_015299.2 Intron NP_056114.1
XM_005267474.4 Intron XP_005267531.1
XM_011536590.2 Intron XP_011534892.1
XM_017021114.1 Intron XP_016876603.1
Gene
NYNRIN
Gene Name
NYN domain and retroviral integrase containing
There are no transcripts associated with this gene.

View Full Product Details