Product Details

SNP ID
rs11206833
Assay Type
Functionally tested
NCBI dbSNP Submissions
55
Location
Chr.1:56499329 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGATACCAAAGTAATTACTGGGCC[C/T]AGAGAAAGCCATGCTCTCCCGTGTG
Phenotype
MIM: 607125
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLPP3 PubMed Links

Gene Details

Gene
PLPP3
Gene Name
phospholipid phosphatase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003713.4 Intron NP_003704.3

View Full Product Details