Product Details

SNP ID
rs11465425
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:7740097 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGGCTGGGGCAGATTTTATATACA[A/G]AGGGTAGTGAGGCATGATATGATTG
Phenotype
MIM: 604672 MIM: 616256
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CD209 PubMed Links

Gene Details

Gene
CD209
Gene Name
CD209 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144893.1 3841 UTR 3 NP_001138365.1
NM_001144894.1 3841 UTR 3 NP_001138366.1
NM_001144895.1 3841 UTR 3 NP_001138367.1
NM_001144896.1 3841 UTR 3 NP_001138368.1
NM_001144897.1 3841 UTR 3 NP_001138369.1
NM_001144899.1 3841 UTR 3 NP_001138371.1
NM_021155.3 3841 UTR 3 NP_066978.1
XM_005272472.4 3841 UTR 3 XP_005272529.1
Gene
CLEC4G
Gene Name
C-type lectin domain family 4 member G
There are no transcripts associated with this gene.

View Full Product Details