Product Details
- SNP ID
-
rs11542895
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:135933361 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTGCCCGGTCTCGGGCCGCACCAGG[G/T]GGCTGCTGTGGCCTGATAGCCGAGT
- Phenotype
-
MIM: 608129
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
UBAC1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115120233,rs151159711] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- UBAC1
- Gene Name
- UBA domain containing 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_016172.2 |
1475 |
UTR 3 |
|
|
NP_057256.2 |
View Full Product Details