Product Details

SNP ID
rs11555750
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100059251 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTAGGAAAAGTAAAAAATTAAAC[C/T]GTTAGCTAAATTCTTCATGGTTTAT
Phenotype
MIM: 194510 MIM: 601261
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF3 PubMed Links

Gene Details

Gene
ZNF3
Gene Name
zinc finger protein 3
There are no transcripts associated with this gene.

Gene
ZSCAN21
Gene Name
zinc finger and SCAN domain containing 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145914.2 Intron NP_666019.1
XM_005250568.4 Intron XP_005250625.1
XM_006716113.3 Intron XP_006716176.1
XM_017012584.1 Intron XP_016868073.1
XM_017012585.1 Intron XP_016868074.1

View Full Product Details