Product Details

SNP ID
rs11555448
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:42090667 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCGCGCACCCTCCGCTTGGCCTC[G/A]TTCATGTCCCGACTGAAAATGGGCT
Phenotype
MIM: 602138 MIM: 615588
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
NDUFA6 PubMed Links

Gene Details

Gene
NDUFA6
Gene Name
NADH:ubiquinone oxidoreductase subunit A6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002490.3 218 Silent Mutation NP_002481.2
Gene
NDUFA6-AS1
Gene Name
NDUFA6 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

Gene
SMDT1
Gene Name
single-pass membrane protein with aspartate rich tail 1
There are no transcripts associated with this gene.

View Full Product Details