Product Details

SNP ID
rs11558135
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:42682654 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCGCCGACACCAAGCCCGGCACTA[C/T]GGGCAGCGGCGCAGGGAGCGGTGGC
Phenotype
MIM: 606095 MIM: 154030
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PPIH PubMed Links

Gene Details

Gene
PPIH
Gene Name
peptidylprolyl isomerase H
There are no transcripts associated with this gene.

Gene
YBX1
Gene Name
Y-box binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004559.4 420 Missense Mutation ACG,ATG T30M NP_004550.2

View Full Product Details