Product Details

SNP ID
rs11608645
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:51257079 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAGAATATAAATAAATGCTTACA[C/T]AAAATTTCTCTGTAAAGGTGAGTAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SMAGP PubMed Links

Gene Details

Gene
SMAGP
Gene Name
small cell adhesion glycoprotein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031628.1 Intron NP_001026798.1
NM_001033873.1 Intron NP_001029045.1

View Full Product Details