Product Details

SNP ID
rs15123
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:106940814 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTGAAAATCTGAAAAAGGCAGGAA[G/T]ATAGGAAGAAGAAAAATATCAACCA
Phenotype
MIM: 300520 MIM: 300970
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CLDN2 PubMed Links

Gene Details

Gene
CLDN2
Gene Name
claudin 2
There are no transcripts associated with this gene.

Gene
MORC4
Gene Name
MORC family CW-type zinc finger 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001085354.2 3716 UTR 3 NP_001078823.1
NM_024657.4 3716 UTR 3 NP_078933.3
XM_005262190.3 3716 UTR 3 XP_005262247.1
XM_006724691.2 3716 UTR 3 XP_006724754.1
XM_011531027.2 3716 UTR 3 XP_011529329.1
XM_017029844.1 3716 UTR 3 XP_016885333.1

View Full Product Details