Product Details
- SNP ID
-
rs180701365
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:151183592 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCTTCTCCAGGGCCTCACGCGTGA[C/G]AAGCAGGTCTTCTGACAGCTGCCTG
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CCDC69
PubMed Links
Gene Details
- Gene
- CCDC69
- Gene Name
- coiled-coil domain containing 69
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_015621.2 |
859 |
Missense Mutation |
CTC,GTC |
L246V |
NP_056436.2 |
View Full Product Details