Product Details

SNP ID
rs180778462
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:14644415 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGAGATGAACCTCTCAAAAGCAC[C/T]GGCCCTCAGATTTCTGTTAGTGAAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BFAR PubMed Links

Gene Details

Gene
BFAR
Gene Name
bifunctional apoptosis regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016561.2 330 Silent Mutation ACC,ACT T23T NP_057645.1
XM_005255350.2 330 UTR 5 XP_005255407.1
XM_011522520.1 330 Silent Mutation ACC,ACT T23T XP_011520822.1

View Full Product Details