Product Details

SNP ID
rs181100025
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:177366277 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGTCCCAGCAGCCCCTTCCCAAC[C/T]GAGGAGCAGCCTGTGGCCAGCTGGG
Phenotype
MIM: 602513
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RGS14 PubMed Links

Gene Details

Gene
RGS14
Gene Name
regulator of G-protein signaling 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006480.4 517 Silent Mutation ACC,ACT T56T NP_006471.2
XM_005265794.4 517 Silent Mutation ACC,ACT T56T XP_005265851.1
XM_005265795.4 517 Silent Mutation ACC,ACT T56T XP_005265852.1

View Full Product Details