Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001015887.1 | 1436 | Missense Mutation | NP_001015887.1 | ||
NM_152538.2 | 1436 | Missense Mutation | NP_689751.2 | ||
XM_006713517.1 | 1436 | Missense Mutation | XP_006713580.1 | ||
XM_011512462.2 | 1436 | Missense Mutation | XP_011510764.1 | ||
XM_011512463.1 | 1436 | Missense Mutation | XP_011510765.1 | ||
XM_011512464.2 | 1436 | Missense Mutation | XP_011510766.1 | ||
XM_011512465.1 | 1436 | Missense Mutation | XP_011510767.1 | ||
XM_011512466.2 | 1436 | Missense Mutation | XP_011510768.1 | ||
XM_011512467.2 | 1436 | Missense Mutation | XP_011510769.1 | ||
XM_011512468.1 | 1436 | Missense Mutation | XP_011510770.1 | ||
XM_011512469.1 | 1436 | Missense Mutation | XP_011510771.1 | ||
XM_011512470.1 | 1436 | Missense Mutation | XP_011510772.1 | ||
XM_011512471.2 | 1436 | Missense Mutation | XP_011510773.1 | ||
XM_011512472.1 | 1436 | Missense Mutation | XP_011510774.1 | ||
XM_011512474.2 | 1436 | Intron | XP_011510776.1 | ||
XM_017005789.1 | 1436 | Missense Mutation | XP_016861278.1 | ||
XM_017005790.1 | 1436 | Missense Mutation | XP_016861279.1 |