Product Details

SNP ID
rs180907329
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:9528926 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTGAATAAGGCGTGAGGATGTAAG[A/G]AAGGATTTCCCACATTCCTTACATT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101928238 PubMed Links

Gene Details

Gene
LOC101928238
Gene Name
uncharacterized LOC101928238
There are no transcripts associated with this gene.

Gene
ZNF426
Gene Name
zinc finger protein 426
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300883.2 1337 Silent Mutation TTC,TTT F335F NP_001287812.1
NM_001318055.1 1337 Intron NP_001304984.1
NM_001318056.1 1337 Intron NP_001304985.1
NM_024106.2 1337 Intron NP_077011.1
XM_017027293.1 1337 Silent Mutation TTC,TTT F335F XP_016882782.1
XM_017027294.1 1337 Silent Mutation TTC,TTT F131F XP_016882783.1

View Full Product Details