Product Details
- SNP ID
-
rs180977020
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:31083256 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGAACCGACGGAGCTAGGGGCCAGC[A/G]AGATGGCGGACGAGGCCTTAGCTGG
- Phenotype
-
MIM: 602127
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SMTN
PubMed Links
Gene Details
- Gene
- SMTN
- Gene Name
- smoothelin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001207017.1 |
323 |
Missense Mutation |
AAG,GAG |
K54E |
NP_001193946.1 |
NM_001207018.1 |
323 |
Intron |
|
|
NP_001193947.1 |
NM_006932.4 |
323 |
UTR 5 |
|
|
NP_008863.3 |
NM_134269.2 |
323 |
UTR 5 |
|
|
NP_599031.1 |
NM_134270.2 |
323 |
UTR 5 |
|
|
NP_599032.2 |
XM_006724298.1 |
323 |
UTR 5 |
|
|
XP_006724361.1 |
XM_006724302.1 |
323 |
UTR 5 |
|
|
XP_006724365.1 |
XM_011530333.2 |
323 |
Missense Mutation |
AAG,GAG |
K54E |
XP_011528635.1 |
XM_011530336.2 |
323 |
Missense Mutation |
AAG,GAG |
K54E |
XP_011528638.1 |
XM_011530337.2 |
323 |
Missense Mutation |
AAG,GAG |
K54E |
XP_011528639.1 |
XM_011530338.2 |
323 |
Missense Mutation |
AAG,GAG |
K54E |
XP_011528640.1 |
XM_011530339.2 |
323 |
Missense Mutation |
AAG,GAG |
K54E |
XP_011528641.1 |
XM_011530340.2 |
323 |
Missense Mutation |
AAG,GAG |
K54E |
XP_011528642.1 |
XM_011530341.1 |
323 |
Intron |
|
|
XP_011528643.1 |
XM_017028913.1 |
323 |
Missense Mutation |
AAG,GAG |
K54E |
XP_016884402.1 |
XM_017028914.1 |
323 |
Missense Mutation |
AAG,GAG |
K54E |
XP_016884403.1 |
XM_017028915.1 |
323 |
Missense Mutation |
AAG,GAG |
K54E |
XP_016884404.1 |
XM_017028916.1 |
323 |
Missense Mutation |
AAG,GAG |
K54E |
XP_016884405.1 |
XM_017028917.1 |
323 |
Missense Mutation |
AAG,GAG |
K54E |
XP_016884406.1 |
XM_017028918.1 |
323 |
Missense Mutation |
AAG,GAG |
K54E |
XP_016884407.1 |
XM_017028919.1 |
323 |
Missense Mutation |
AAG,GAG |
K54E |
XP_016884408.1 |
View Full Product Details