Product Details

SNP ID
rs181001236
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:45688448 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTTGCGGCAGTTGATGAGCACTT[G/T]GGTATTGTTCTTGACTGACTGTGTG
Phenotype
MIM: 137241 MIM: 601061
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GIPR PubMed Links

Gene Details

Gene
GIPR
Gene Name
gastric inhibitory polypeptide receptor
There are no transcripts associated with this gene.

Gene
QPCTL
Gene Name
glutaminyl-peptide cyclotransferase like
There are no transcripts associated with this gene.

Gene
SNRPD2
Gene Name
small nuclear ribonucleoprotein D2 polypeptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004597.6 Intron NP_004588.1
NM_177542.3 Intron NP_808210.2

View Full Product Details